Learn More
Medchemexpress LLC DBCO-Biotin | 1418217-95-4 | MFCD22380751 | 98.5% | 502.63 g/mol | C28H30N4O3S | 100MG
SDP
Catalog No. 5000236792
Encompass_Preferred
DBCO-Biotin is a dibenzocyclooctyne-linked biotin reagent designed for copper-free click chemistry and efficient biotinylation of azide-functionalized molecules. It is used for catalyst-free conjugation in labeling, enrichment, and linker assembly workflows, including PROTAC synthesis. The reagent is DMSO-soluble and supplied with recommended storage conditions to preserve stability.
Catalog No. 50-002-36792 Supplier Medchemexpress LLC Supplier No. HY123916100MG
DBCO-Biotin is a dibenzocyclooctyne-linked biotin reagent designed for copper-free click chemistry and efficient biotinylation of azide-functionalized molecules. It is used for catalyst-free conjugation in labeling, enrichment, and linker assembly workflows, including PROTAC synthesis. The reagent is DMSO-soluble and supplied with recommended storage conditions to preserve stability.