Learn More
Medchemexpress LLC Dbco-biotin | 1418217-95-4 | MFCD22380751 | 98.5% | 502.63 g/mol | C28H30N4O3S | 25MG
SDP
Catalog No. 5000236991
Encompass_Preferred
DBCO-Biotin is a copper-free click chemistry reagent that installs a biotin tag on azide-functionalized biomolecules via strain-promoted alkyne-azide cycloaddition (SPAAC). Supplied as a white powder, it is used for bioconjugation, affinity purification and detection, and as a linker component in targeted degrader synthesis. Store at -20 °C, protect from light, and maintain under inert atmosphere when possible.
  • Enables copper-free SPAAC bioconjugation to azide-labeled targets.
  • Introduces a biotin affinity handle for pull-down and detection assays.
  • High purity suitable for analytical and preparative applications.
  • Stable solid form when stored at -20 °C and protected from light.
  • Compatible with proteins, peptides, and other biomolecules.
Catalog No. 50-002-36991 Supplier Medchemexpress LLC Supplier No. HY12391625MG
DBCO-Biotin is a copper-free click chemistry reagent that installs a biotin tag on azide-functionalized biomolecules via strain-promoted alkyne-azide cycloaddition (SPAAC). Supplied as a white powder, it is used for bioconjugation, affinity purification and detection, and as a linker component in targeted degrader synthesis. Store at -20 °C, protect from light, and maintain under inert atmosphere when possible.
  • Enables copper-free SPAAC bioconjugation to azide-labeled targets.
  • Introduces a biotin affinity handle for pull-down and detection assays.
  • High purity suitable for analytical and preparative applications.
  • Stable solid form when stored at -20 °C and protected from light.
  • Compatible with proteins, peptides, and other biomolecules.