Learn More
Medchemexpress LLC DBCO-biotin | 1418217-95-4 | MFCD22380751 | 98.5% | 502.63 g·mol⁻¹ | C28H30N4O3S | 5MG
SDP
Catalog No. 5000237983
Encompass_Preferred
DBCO-biotin is a dibenzocyclooctyne (DBCO)-linked biotin reagent designed for copper-free, bioorthogonal click chemistry. It enables rapid biotinylation of azide-functionalized molecules for affinity capture, labeling, and conjugation workflows, and is suitable as a linker component in PROTAC and bioconjugation applications.
  • Enables copper-free click conjugation to azide-containing molecules.
  • Provides a biotin tag for affinity capture and detection.
  • High reported purity for consistent experimental performance.
  • Available in small research quantities for method development.
  • Solid, easy-to-handle form with standard storage recommendations.
Catalog No. 50-002-37983 Supplier Medchemexpress LLC Supplier No. HY1239165MG
DBCO-biotin is a dibenzocyclooctyne (DBCO)-linked biotin reagent designed for copper-free, bioorthogonal click chemistry. It enables rapid biotinylation of azide-functionalized molecules for affinity capture, labeling, and conjugation workflows, and is suitable as a linker component in PROTAC and bioconjugation applications.
  • Enables copper-free click conjugation to azide-containing molecules.
  • Provides a biotin tag for affinity capture and detection.
  • High reported purity for consistent experimental performance.
  • Available in small research quantities for method development.
  • Solid, easy-to-handle form with standard storage recommendations.