Learn More
Medchemexpress LLC Dbco-c3-acid | 1207355-31-4 | 99.7% | 319.36 g/mol | C20H17NO3 | 250 MG
SDP
Catalog No. 5000204043
Encompass_Preferred
DBCO-C3-Acid is a dibenzylcyclooctyne-functionalized carboxylic acid used as a click-chemistry intermediate for assembling antibody-drug conjugate linkers and other bioconjugates. It supplies a strained alkyne handle for copper-free strain-promoted azide-alkyne cycloaddition (SPAAC), enabling efficient coupling to azide-containing partners. The compound is supplied as a solid with documented purity and recommended storage conditions.
  • Used as a click-chemistry intermediate for ADC linker and bioconjugate synthesis.
  • Provides a strained alkyne handle for copper-free SPAAC with azides.
  • High purity of 99.7% as reported by manufacturer.
  • Molecular weight 319.36 g/mol; formula C20H17NO3.
  • Stable as a powder when stored under recommended temperatures.
Catalog No. 50-002-04043 Supplier Medchemexpress LLC Supplier No. HY120903250MG
DBCO-C3-Acid is a dibenzylcyclooctyne-functionalized carboxylic acid used as a click-chemistry intermediate for assembling antibody-drug conjugate linkers and other bioconjugates. It supplies a strained alkyne handle for copper-free strain-promoted azide-alkyne cycloaddition (SPAAC), enabling efficient coupling to azide-containing partners. The compound is supplied as a solid with documented purity and recommended storage conditions.
  • Used as a click-chemistry intermediate for ADC linker and bioconjugate synthesis.
  • Provides a strained alkyne handle for copper-free SPAAC with azides.
  • High purity of 99.7% as reported by manufacturer.
  • Molecular weight 319.36 g/mol; formula C20H17NO3.
  • Stable as a powder when stored under recommended temperatures.