Learn More
Medchemexpress LLC DBCO-C3-amide-PEG6-NHS ester | 95.2% | 751.82 | 1 MG
SDP
Catalog No. 5000393021
Encompass_Preferred
DBCO-C3-amide-PEG6-NHS ester is a click chemistry PEG reagent containing an NHS ester that reacts specifically with primary amines, such as side chains of lysine residues or aminosilane-coated surfaces, under neutral or slightly alkaline conditions, to form efficient covalent bonds. It also contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • Click chemistry PEG reagent
  • Contains an NHS ester that reacts specifically with primary amines
  • Reacts under neutral or slightly alkaline conditions
  • Forms efficient covalent bonds
  • Contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC)
Catalog No. 50-003-93021 Supplier Medchemexpress LLC Supplier No. HY160773A1MG
DBCO-C3-amide-PEG6-NHS ester is a click chemistry PEG reagent containing an NHS ester that reacts specifically with primary amines, such as side chains of lysine residues or aminosilane-coated surfaces, under neutral or slightly alkaline conditions, to form efficient covalent bonds. It also contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • Click chemistry PEG reagent
  • Contains an NHS ester that reacts specifically with primary amines
  • Reacts under neutral or slightly alkaline conditions
  • Forms efficient covalent bonds
  • Contains a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC)