Learn More
Medchemexpress LLC DBCO-C3-amide-PEG6-NHS ester | 95.2% | 751.82 | 5 MG
SDP
Catalog No. 5000393069
Encompass_Preferred
DBCO-C3-amide-PEG6-NHS ester is a click chemistry PEG reagent. It contains an NHS ester that reacts specifically with primary amines, such as the side chains of lysine residues or aminosilane-coated surfaces, under neutral or slightly alkaline conditions to form covalent bonds. It also contains a DBCO group which can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • Contains an NHS ester that reacts with primary amines
  • Forms covalent bonds under neutral or slightly alkaline conditions
  • Features a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Suitable for reactions with molecules containing azide groups
Catalog No. 50-003-93069 Supplier Medchemexpress LLC Supplier No. HY160773A5MG
DBCO-C3-amide-PEG6-NHS ester is a click chemistry PEG reagent. It contains an NHS ester that reacts specifically with primary amines, such as the side chains of lysine residues or aminosilane-coated surfaces, under neutral or slightly alkaline conditions to form covalent bonds. It also contains a DBCO group which can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • Contains an NHS ester that reacts with primary amines
  • Forms covalent bonds under neutral or slightly alkaline conditions
  • Features a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Suitable for reactions with molecules containing azide groups