Learn More
Medchemexpress LLC Dbco-C6-acid | 1425485-72-8 | 98.9% | 333.38 g·mol⁻¹ | C21H19NO3 | 1 G
SDP
Catalog No. 5000202765
Encompass_Preferred
DBCO-C6-acid is a dibenzocyclooctyne-containing linker used for click chemistry and antibody-drug conjugate assembly. It contains a strained alkyne that reacts with azide groups via strain-promoted alkyne-azide cycloaddition (SPAAC), enabling copper-free bioconjugation workflows in organic synthesis and bioconjugation applications.
  • Enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Serves as a non-cleavable linker for antibody-drug conjugate assembly.
  • Supplied as a high-purity solid suitable for organic synthesis and bioconjugation.
  • Stable when stored under recommended conditions for long-term use.
  • Available in multiple pack sizes to support small- and larger-scale workflows.
Catalog No. 50-002-02765 Supplier Medchemexpress LLC Supplier No. HY1218051G
Add to Cart
Add to Cart
DBCO-C6-acid is a dibenzocyclooctyne-containing linker used for click chemistry and antibody-drug conjugate assembly. It contains a strained alkyne that reacts with azide groups via strain-promoted alkyne-azide cycloaddition (SPAAC), enabling copper-free bioconjugation workflows in organic synthesis and bioconjugation applications.
  • Enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • Serves as a non-cleavable linker for antibody-drug conjugate assembly.
  • Supplied as a high-purity solid suitable for organic synthesis and bioconjugation.
  • Stable when stored under recommended conditions for long-term use.
  • Available in multiple pack sizes to support small- and larger-scale workflows.