Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbco-conh-s-s-nhs ester | 1435934-53-4 | MFCD22380758 | 99.2% | 565.66 g/mol | C28H27N3O6S2 | 5 MG
SDP

Catalog No. 5000202574
Encompass_Preferred

DBCO-CONH-S-S-NHS ester is a cleavable disulfide-containing ADC linker that combines a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with an N-hydroxysuccinimide (NHS) activated ester for conjugation to amines.

  • Provides a cleavable disulfide spacer for controlled payload release.
  • Enables copper-free click chemistry via DBCO for bioorthogonal conjugation.
  • Contains an NHS-activated ester for efficient amine coupling.
  • High reported purity supports sensitive bioconjugation workflows.
  • Soluble in DMSO at high concentrations for easy handling and stock preparation.
  • Stable when stored protected from light and under inert atmosphere at low temperatures.

Catalog No. 50-002-02574 Supplier Medchemexpress LLC Supplier No. HY1334135MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-CONH-S-S-NHS ester is a cleavable disulfide-containing ADC linker that combines a DBCO group for strain-promoted alkyne-azide cycloaddition (SPAAC) with an N-hydroxysuccinimide (NHS) activated ester for conjugation to amines.

  • Provides a cleavable disulfide spacer for controlled payload release.
  • Enables copper-free click chemistry via DBCO for bioorthogonal conjugation.
  • Contains an NHS-activated ester for efficient amine coupling.
  • High reported purity supports sensitive bioconjugation workflows.
  • Soluble in DMSO at high concentrations for easy handling and stock preparation.
  • Stable when stored protected from light and under inert atmosphere at low temperatures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.