Learn More
Medchemexpress LLC DBCO-dextran sulfate | 98.4% | 40,000 (average) | 10 MG
SDP
Catalog No. 5000289011
Encompass_Preferred
DBCO-dextran sulfate (average molecular weight 40,000) is a DBCO-functionalized dextran sulfate supplied as a powder for research use. It is used to construct DS-EXOs in cells via strain-promoted azide-alkyne click chemistry (SPAAC), and is applicable to studies such as rheumatoid arthritis research. Store powder at -20°C; in solution, store at -80°C for up to 6 months or -20°C for up to 1 month.
  • DBCO-functionalized for copper-free click chemistry
  • Average molecular weight 40,000
  • High purity (HPLC 98.35%)
  • Supplied as a powder for long-term storage stability
  • Suitable for constructing DS-EXOs in cellular research models
Catalog No. 50-002-89011 Supplier Medchemexpress LLC Supplier No. HY16441610MG
DBCO-dextran sulfate (average molecular weight 40,000) is a DBCO-functionalized dextran sulfate supplied as a powder for research use. It is used to construct DS-EXOs in cells via strain-promoted azide-alkyne click chemistry (SPAAC), and is applicable to studies such as rheumatoid arthritis research. Store powder at -20°C; in solution, store at -80°C for up to 6 months or -20°C for up to 1 month.
  • DBCO-functionalized for copper-free click chemistry
  • Average molecular weight 40,000
  • High purity (HPLC 98.35%)
  • Supplied as a powder for long-term storage stability
  • Suitable for constructing DS-EXOs in cellular research models