Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-β-Glu-PEG12-Exatecan | 3025107-83-6 | 98.6% | 1833.95 | 1 MG
SDP

Catalog No. 5000394145
Encompass_Preferred

DBCO-β-Glu-PEG12-Exatecan is an inhibitor for topoisomerase I and can be utilized as a Drug-Linker Conjugate for ADC molecules.

  • Inhibitor for topoisomerase I
  • Drug-Linker Conjugate for ADC molecules
  • Purity of 98.63%
  • Solid appearance, off-white to light yellow color
  • Recommended storage at -20°C (sealed, away from moisture and light)
  • Extended storage in solvent at -80°C for 6 months, or -20°C for 1 month

Catalog No. 50-003-94145 Supplier Medchemexpress LLC Supplier No. HY1607881MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-β-Glu-PEG12-Exatecan is an inhibitor for topoisomerase I and can be utilized as a Drug-Linker Conjugate for ADC molecules.

  • Inhibitor for topoisomerase I
  • Drug-Linker Conjugate for ADC molecules
  • Purity of 98.63%
  • Solid appearance, off-white to light yellow color
  • Recommended storage at -20°C (sealed, away from moisture and light)
  • Extended storage in solvent at -80°C for 6 months, or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.