Learn More
Medchemexpress LLC DBCO-β-Glu-PEG12-Exatecan | 3025107-83-6 | 98.6% | 1833.95 | 5 MG
SDP
Catalog No. 5000394366
Encompass_Preferred
DBCO-β-Glu-PEG12-Exatecan is an inhibitor for topoisomerase I and can be used as a Drug-Linker Conjugate for ADC molecules.
  • Acts as an inhibitor for topoisomerase I.
  • Can be utilized as a drug-linker conjugate for antibody-drug conjugate (ADC) molecules.
Catalog No. 50-003-94366 Supplier Medchemexpress LLC Supplier No. HY1607885MG
DBCO-β-Glu-PEG12-Exatecan is an inhibitor for topoisomerase I and can be used as a Drug-Linker Conjugate for ADC molecules.
  • Acts as an inhibitor for topoisomerase I.
  • Can be utilized as a drug-linker conjugate for antibody-drug conjugate (ADC) molecules.