Learn More
Medchemexpress LLC DBCO-mPEG | 00-00-0 | 98.4% | 10MG
SDP
Catalog No. 5000261471
Encompass_Preferred
DBCO-mPEG is a DBCO-functionalized methoxy-polyethylene glycol (mPEG) reagent with an average molecular weight of 5,000 Da. Supplied as a white to light yellow solid for research use, it enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) to conjugate azide-containing molecules and serves as a spacer or linker in bioconjugation and PROTAC synthesis.
  • Enables copper-free click chemistry (SPAAC) with azide-containing partners.
  • Provides a methoxy-terminated PEG spacer with average MW 5,000 Da.
  • Supplied as a solid with high reported purity (~98.4%).
  • Suitable for bioconjugation, linker construction, and PROTAC applications.
  • Stable when stored at -20°C under inert atmosphere; in solution, store at -80°C for long-term.
Catalog No. 50-002-61471 Supplier Medchemexpress LLC Supplier No. HY14031910MG
DBCO-mPEG is a DBCO-functionalized methoxy-polyethylene glycol (mPEG) reagent with an average molecular weight of 5,000 Da. Supplied as a white to light yellow solid for research use, it enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) to conjugate azide-containing molecules and serves as a spacer or linker in bioconjugation and PROTAC synthesis.
  • Enables copper-free click chemistry (SPAAC) with azide-containing partners.
  • Provides a methoxy-terminated PEG spacer with average MW 5,000 Da.
  • Supplied as a solid with high reported purity (~98.4%).
  • Suitable for bioconjugation, linker construction, and PROTAC applications.
  • Stable when stored at -20°C under inert atmosphere; in solution, store at -80°C for long-term.