Learn More
Medchemexpress LLC DBCO-mPEG (MW 30kDa) | 90.0% | 25 MG
SDP
Catalog No. 5000352570
Encompass_Preferred
DBCO-mPEG (MW 30kDa) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Features a DBCO group
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-52570 Supplier Medchemexpress LLC Supplier No. 25MGHY140322
DBCO-mPEG (MW 30kDa) is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Functions as a click chemistry reagent
  • Features a DBCO group
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules
  • Used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system to selectively degrade target proteins