Learn More
Medchemexpress LLC DBCO-mPEG (MW 5kDa) | 99.1% | 25 MG
SDP
Catalog No. 5000352577
Encompass_Preferred
DBCO-mPEG (MW 5kDa) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group capable of strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs utilize the intracellular ubiquitin-proteasome system for the selective degradation of target proteins by connecting an E3 ubiquitin ligase ligand and a specific protein target ligand with a linker.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC with azide groups
  • Used in the synthesis of PROTACs
  • Leverages the ubiquitin-proteasome system for selective protein degradation
Catalog No. 50-003-52577 Supplier Medchemexpress LLC Supplier No. 25MGHY140319
DBCO-mPEG (MW 5kDa) is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group capable of strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups. PROTACs utilize the intracellular ubiquitin-proteasome system for the selective degradation of target proteins by connecting an E3 ubiquitin ligase ligand and a specific protein target ligand with a linker.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC with azide groups
  • Used in the synthesis of PROTACs
  • Leverages the ubiquitin-proteasome system for selective protein degradation