Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-N-bis(PEG4-NHS ester) | 2639395-38-1 | 96.2% | 995.03 | C49H62N4O18 | 5 MG
SDP

Catalog No. 5000203686
Encompass_Preferred

DBCO-N-bis(PEG4-NHS ester) is a PEG-based bifunctional linker that contains two PEG4-NHS ester groups and a dibenzocyclooctyne (DBCO) moiety. It is designed for bioconjugation and protein labeling via NHS-ester chemistry and for copper-free click conjugation with azide-containing partners through strain-promoted alkyne-azide cycloaddition (SPAAC).

  • Contains two PEG4-NHS ester groups and a DBCO functional group.
  • Enables protein modification through NHS-ester conjugation to primary amines.
  • Supports copper-free SPAAC click chemistry with azide partners.
  • Suitable for small-scale research use; available in 5 mg and larger sizes.
  • Molecular weight 995.03; CAS number 2639395-38-1.

Catalog No. 50-002-03686 Supplier Medchemexpress LLC Supplier No. HY1450905MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-N-bis(PEG4-NHS ester) is a PEG-based bifunctional linker that contains two PEG4-NHS ester groups and a dibenzocyclooctyne (DBCO) moiety. It is designed for bioconjugation and protein labeling via NHS-ester chemistry and for copper-free click conjugation with azide-containing partners through strain-promoted alkyne-azide cycloaddition (SPAAC).

  • Contains two PEG4-NHS ester groups and a DBCO functional group.
  • Enables protein modification through NHS-ester conjugation to primary amines.
  • Supports copper-free SPAAC click chemistry with azide partners.
  • Suitable for small-scale research use; available in 5 mg and larger sizes.
  • Molecular weight 995.03; CAS number 2639395-38-1.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.