Learn More
Medchemexpress LLC Dbco-nh-boc | 1539290-74-8 | C23H24N2O3 | 100 MG
SDP
Catalog No. 5000357313
Encompass_Preferred
DBCO-NH-Boc is an alkyl/ether-based PROTAC linker. It acts as a click chemistry reagent due to its DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs are composed of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
  • Alkyl/ether-based PROTAC linker
  • Click chemistry reagent
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used in the synthesis of PROTACs
Catalog No. 50-003-57313 Supplier Medchemexpress LLC Supplier No. HY140293100MG
DBCO-NH-Boc is an alkyl/ether-based PROTAC linker. It acts as a click chemistry reagent due to its DBCO group, which facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs are composed of two different ligands connected by a linker, one for an E3 ubiquitin ligase and the other for the target protein, utilizing the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is intended for research use only.
  • Alkyl/ether-based PROTAC linker
  • Click chemistry reagent
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used in the synthesis of PROTACs