Learn More
Medchemexpress LLC DBCO-NHCO-PEG2-biotin | 2989400-08-8 | 97.1% | C35H43N5O6S | 10MG
SDP
Catalog No. 5000259192
Encompass_Preferred
DBCO-NHCO-PEG2-Biotin is a PEG-based click chemistry linker with a dibenzocyclooctyne (DBCO) reactive group and a PEG2 spacer, designed for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC). It is intended for bioconjugation, labeling, affinity capture, and linker assembly in chemical biology and PROTAC research.
  • Enables copper-free SPAAC with azide-functionalized partners.
  • Provides biotin affinity for streptavidin/avidin capture.
  • Contains a PEG2 spacer for flexibility and improved solubility.
  • High purity suitable for conjugation and analytical workflows.
  • Available in small research-scale quantities for laboratory use.
Catalog No. 50-002-59192 Supplier Medchemexpress LLC Supplier No. HY13593910MG
DBCO-NHCO-PEG2-Biotin is a PEG-based click chemistry linker with a dibenzocyclooctyne (DBCO) reactive group and a PEG2 spacer, designed for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC). It is intended for bioconjugation, labeling, affinity capture, and linker assembly in chemical biology and PROTAC research.
  • Enables copper-free SPAAC with azide-functionalized partners.
  • Provides biotin affinity for streptavidin/avidin capture.
  • Contains a PEG2 spacer for flexibility and improved solubility.
  • High purity suitable for conjugation and analytical workflows.
  • Available in small research-scale quantities for laboratory use.