Learn More
Medchemexpress LLC Dbco-nhco-peg2-maleimide | 2698339-31-8 | MFCD31747082 | 97.7% | 586.63 | C32H34N4O7 | 5 MG
SDP
Catalog No. 5000264925
Encompass_Preferred
DBCO-NHCO-PEG2-maleimide is a heterobifunctional PEG-based linker for bioconjugation that combines a strained alkyne (DBCO) with a maleimide group to enable copper-free click chemistry and thiol-specific coupling between azide- and thiol-containing molecules in research applications.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azides.
  • Provides maleimide-mediated thiol conjugation for cysteine residues.
  • PEG2 spacer improves solubility and reduces steric hindrance.
  • High purity (~97.7%) suitable for research use.
  • Yellow to brown solid that is stable when stored at -20°C.
  • Molecular weight ~586.63 and formula C32H34N4O7 for reference.
Catalog No. 50-002-64925 Supplier Medchemexpress LLC Supplier No. HY1333725MG
DBCO-NHCO-PEG2-maleimide is a heterobifunctional PEG-based linker for bioconjugation that combines a strained alkyne (DBCO) with a maleimide group to enable copper-free click chemistry and thiol-specific coupling between azide- and thiol-containing molecules in research applications.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azides.
  • Provides maleimide-mediated thiol conjugation for cysteine residues.
  • PEG2 spacer improves solubility and reduces steric hindrance.
  • High purity (~97.7%) suitable for research use.
  • Yellow to brown solid that is stable when stored at -20°C.
  • Molecular weight ~586.63 and formula C32H34N4O7 for reference.