Learn More
Medchemexpress LLC Dbco-nhco-peg3-fmoc | 00-00-0 | 96.9% | C42H43N3O7 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13347210MG
Encompass_Preferred
DBCO-NHCO-PEG3-Fmoc is a DBCO-functionalized PEG3 linker with an Fmoc-protected amine designed for use as a click-chemistry reagent in PROTAC synthesis and bioconjugation. It participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing partners, offering modular attachment and improved handling for linker assembly.
  • Click-reactive DBCO group enables strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Peg3 spacer improves solubility and reduces steric hindrance.
  • Fmoc-protected amine allows compatibility with solid-phase and solution-phase synthesis workflows.
  • High purity suitable for research applications.
  • Stable as powder at -20°C for extended storage and recommended cold storage in solution.
Catalog No. 50-002-59483