Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbco-nhco-peg3-fmoc | 96.9% | 701.81 g/mol | C42H43N3O7 | 5 MG
SDP

Catalog No. 5000263035
Encompass_Preferred

DBCO-NHCO-PEG3-Fmoc is a PEG-based linker containing a dibenzocyclooctyne (DBCO) moiety and an Fmoc-protected amine. It is designed for strain-promoted alkyne-azide cycloaddition (SPAAC) and for use as a linker in PROTAC and conjugation chemistry. Supplied as a light-yellow solid for research applications.

  • Purity 96.9%.
  • Molecular weight 701.81 g/mol.
  • Molecular formula C42H43N3O7.
  • Physical state solid, light yellow to yellow.
  • Storage: powder stable at -20°C for up to 3 years.
  • Applications include PROTAC synthesis and SPAAC click chemistry.

Catalog No. 50-002-63035 Supplier Medchemexpress LLC Supplier No. HY1334725MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-NHCO-PEG3-Fmoc is a PEG-based linker containing a dibenzocyclooctyne (DBCO) moiety and an Fmoc-protected amine. It is designed for strain-promoted alkyne-azide cycloaddition (SPAAC) and for use as a linker in PROTAC and conjugation chemistry. Supplied as a light-yellow solid for research applications.

  • Purity 96.9%.
  • Molecular weight 701.81 g/mol.
  • Molecular formula C42H43N3O7.
  • Physical state solid, light yellow to yellow.
  • Storage: powder stable at -20°C for up to 3 years.
  • Applications include PROTAC synthesis and SPAAC click chemistry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.