Learn More
Medchemexpress LLC Dbco-nhco-peg4-nh-boc | 1255942-12-1 | 623.74 g·mol⁻1 | C34H45N3O8 | 100 MG
SDP
Catalog No. 5000202079
Encompass_Preferred
DBCO-NHCO-PEG4-NH-Boc is a DBCO-functionalized PEG4 linker containing a Boc-protected amine used for copper-free click chemistry, PROTAC and ADC linker synthesis, and general bioconjugation.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free click reactions.
  • PEG4 spacer improves solubility and provides flexible linkage.
  • Boc-protected amine allows orthogonal deprotection and downstream conjugation.
  • Suitable for PROTAC and ADC synthesis and other bioconjugation workflows.
  • Refer to the product datasheet for storage, handling, and lot-specific purity information.
Catalog No. 50-002-02079 Supplier Medchemexpress LLC Supplier No. HY126884100MG
DBCO-NHCO-PEG4-NH-Boc is a DBCO-functionalized PEG4 linker containing a Boc-protected amine used for copper-free click chemistry, PROTAC and ADC linker synthesis, and general bioconjugation.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) for copper-free click reactions.
  • PEG4 spacer improves solubility and provides flexible linkage.
  • Boc-protected amine allows orthogonal deprotection and downstream conjugation.
  • Suitable for PROTAC and ADC synthesis and other bioconjugation workflows.
  • Refer to the product datasheet for storage, handling, and lot-specific purity information.