Learn More
Medchemexpress LLC Dbco-nhco-peg4-nh-boc | 1255942-12-1 | MFCD30730371 | >96.0% | 623.74 g/mol | C34H45N3O8 | 25 MG
SDP
Catalog No. 5000204129
Encompass_Preferred
Dbco-nhco-peg4-nh-boc is a DBCO-functionalized PEG4 linker used for copper-free click chemistry (SPAAC) in bioconjugation, antibody-drug conjugate assembly, and PROTAC synthesis.
  • Enables copper-free click chemistry with azide-functionalized molecules.
  • Suitable for antibody-drug conjugate and PROTAC linker synthesis.
  • Boc-protected amine facilitates selective deprotection and further functionalization.
  • Available in small laboratory quantities for research use.
  • Stable when stored at recommended temperatures.
Catalog No. 50-002-04129 Supplier Medchemexpress LLC Supplier No. HY12688425MG
Dbco-nhco-peg4-nh-boc is a DBCO-functionalized PEG4 linker used for copper-free click chemistry (SPAAC) in bioconjugation, antibody-drug conjugate assembly, and PROTAC synthesis.
  • Enables copper-free click chemistry with azide-functionalized molecules.
  • Suitable for antibody-drug conjugate and PROTAC linker synthesis.
  • Boc-protected amine facilitates selective deprotection and further functionalization.
  • Available in small laboratory quantities for research use.
  • Stable when stored at recommended temperatures.