Learn More
Medchemexpress LLC Dbco-NHCO-PEG4-NH-Boc | 1255942-12-1 | MFCD30730371 | 98.0% | 623.74 g/mol | C34H45N3O8 | 50 MG
SDP
Catalog No. 5000204130
Encompass_Preferred
DBCO-NHCO-PEG4-NH-Boc is a DBCO-containing PEG4-based linker and click chemistry reagent used for strain-promoted alkyne-azide cycloaddition (SPAAC). It is commonly applied in antibody-drug conjugate assembly, PROTAC construction, and site-specific bioconjugation. The molecule contains a Boc-protected amine and a PEG4 spacer to improve solubility and flexibility.
  • enables copper-free SPAAC bioconjugation
  • contains a PEG4 spacer for improved solubility
  • features a Boc-protected amine for orthogonal deprotection
  • suitable for ADC and PROTAC linker synthesis
  • provided as a small-scale reagent for research use
Catalog No. 50-002-04130 Supplier Medchemexpress LLC Supplier No. HY12688450MG
DBCO-NHCO-PEG4-NH-Boc is a DBCO-containing PEG4-based linker and click chemistry reagent used for strain-promoted alkyne-azide cycloaddition (SPAAC). It is commonly applied in antibody-drug conjugate assembly, PROTAC construction, and site-specific bioconjugation. The molecule contains a Boc-protected amine and a PEG4 spacer to improve solubility and flexibility.
  • enables copper-free SPAAC bioconjugation
  • contains a PEG4 spacer for improved solubility
  • features a Boc-protected amine for orthogonal deprotection
  • suitable for ADC and PROTAC linker synthesis
  • provided as a small-scale reagent for research use