Learn More
Medchemexpress LLC Dbco-nhco-peg5-nhs ester | 1378531-80-6 | C36H43N3O11 | 1 MG
SDP
Catalog No. 5000357116
Encompass_Preferred
DBCO-NHCO-PEG5-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules that contain azide groups. PROTACs consist of two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC
  • Exploits intracellular ubiquitin-proteasome system
Catalog No. 50-003-57116 Supplier Medchemexpress LLC Supplier No. HY1402751MG
DBCO-NHCO-PEG5-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group capable of undergoing strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules that contain azide groups. PROTACs consist of two different ligands connected by a linker, where one ligand targets an E3 ubiquitin ligase and the other targets a specific protein, exploiting the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC
  • Exploits intracellular ubiquitin-proteasome system