Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbco-nhco-peg6-biotin | 3081794-36-4 | 96.7% | C43H59N5O10S | 10MG
SDP

Catalog No. 5000260503
Encompass_Preferred

DBCO-NHCO-PEG6-Biotin is a PEG-based bioconjugation linker bearing a strain-promoted dibenzocyclooctyne (DBCO) moiety and a biotin tag. It is designed for copper-free click chemistry to couple azide-containing molecules for detection, enrichment, or incorporation into bifunctional constructs such as PROTACs. Supplied as a colorless to light yellow oil with high purity and DMSO solubility.

  • Enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Provides a biotin tag for affinity enrichment and detection.
  • Contains a PEG6 spacer to improve solubility and reduce steric hindrance.
  • High purity suitable for bioconjugation and linker synthesis.
  • Soluble in DMSO for convenient formulation.
  • Stable when stored under inert atmosphere at recommended temperatures.

Catalog No. 50-002-60503 Supplier Medchemexpress LLC Supplier No. HY13595810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-NHCO-PEG6-Biotin is a PEG-based bioconjugation linker bearing a strain-promoted dibenzocyclooctyne (DBCO) moiety and a biotin tag. It is designed for copper-free click chemistry to couple azide-containing molecules for detection, enrichment, or incorporation into bifunctional constructs such as PROTACs. Supplied as a colorless to light yellow oil with high purity and DMSO solubility.

  • Enables copper-free strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Provides a biotin tag for affinity enrichment and detection.
  • Contains a PEG6 spacer to improve solubility and reduce steric hindrance.
  • High purity suitable for bioconjugation and linker synthesis.
  • Soluble in DMSO for convenient formulation.
  • Stable when stored under inert atmosphere at recommended temperatures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.