Learn More
Broadpharm DBCO-NHS, 1353016-71-3, MFCD24386367, 1g
Catalog No. NC1615784
Encompass
Formula: C23H18N2O5, Purity: 97%, MW: 402.4, DBCO-NHS Ester is an amine-reactive compound and can be used to modify an amine-containing molecule in organic solvents (limited solubility in aqueous media). It reacts with primary amines such as the side chain of lysine residues or aminosilane-coated surfaces at neutral or slightly basic pH to form covalent bonds. The low mass weight will add minimal spacer to modified molecules.
Catalog No. NC1615784 Supplier Broadpharm Supplier No. BP222311G
Formula: C23H18N2O5, Purity: 97%, MW: 402.4, DBCO-NHS Ester is an amine-reactive compound and can be used to modify an amine-containing molecule in organic solvents (limited solubility in aqueous media). It reacts with primary amines such as the side chain of lysine residues or aminosilane-coated surfaces at neutral or slightly basic pH to form covalent bonds. The low mass weight will add minimal spacer to modified molecules.