Learn More
Medchemexpress LLC DBCO-NHS ester | 1353016-71-3 | 99.1% | 402.40 g·mol⁻1 | C23H18N2O5 | 5 MG
SDP
Catalog No. 5000202128
Encompass_Preferred
DBCO-NHS ester is a dibenzocyclooctyne-derived N-hydroxysuccinimide (NHS) ester for copper-free click bioconjugation via strain-promoted alkyne-azide cycloaddition (SPAAC). It is used to install linkers and conjugate azide-bearing molecules in chemical biology and antibody-drug conjugate (ADC) workflows.
  • Enables copper-free SPAAC with azide-containing molecules.
  • Suitable for linker installation and bioconjugation in ADC and biomolecule conjugation workflows.
  • Reported high purity for analytical and preparative use.
  • Available in small research quantities, including 5 MG format.
Catalog No. 50-002-02128 Supplier Medchemexpress LLC Supplier No. HY429735MG
DBCO-NHS ester is a dibenzocyclooctyne-derived N-hydroxysuccinimide (NHS) ester for copper-free click bioconjugation via strain-promoted alkyne-azide cycloaddition (SPAAC). It is used to install linkers and conjugate azide-bearing molecules in chemical biology and antibody-drug conjugate (ADC) workflows.
  • Enables copper-free SPAAC with azide-containing molecules.
  • Suitable for linker installation and bioconjugation in ADC and biomolecule conjugation workflows.
  • Reported high purity for analytical and preparative use.
  • Available in small research quantities, including 5 MG format.