Learn More
Medchemexpress LLC DBCO-NHS ester | 1353016-71-3 | 99.5% | 402.40 | C23H18N2O5 | 10 MG
SDP
Catalog No. 5000202046
Encompass_Preferred
DBCO-NHS ester is a dibenzocyclooctyne N-hydroxysuccinimidyl ester used as a click-chemistry linker to enable strain-promoted alkyne-azide cycloaddition (SPAAC) for bioconjugation and antibody-drug conjugate synthesis. Supplied as a powder with recommended storage and handling guidance; suitable for laboratory research applications.
  • Click-chemistry reagent for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • High purity (≈99.5% by LCMS).
  • Molecular weight 402.40, chemical formula C23H18N2O5.
  • Available in small-mg packages for research use.
  • Store as powder at -20°C; follow SDS for solution storage and handling.
Catalog No. 50-002-02046 Supplier Medchemexpress LLC Supplier No. HY4297310MG
DBCO-NHS ester is a dibenzocyclooctyne N-hydroxysuccinimidyl ester used as a click-chemistry linker to enable strain-promoted alkyne-azide cycloaddition (SPAAC) for bioconjugation and antibody-drug conjugate synthesis. Supplied as a powder with recommended storage and handling guidance; suitable for laboratory research applications.
  • Click-chemistry reagent for strain-promoted alkyne-azide cycloaddition (SPAAC).
  • High purity (≈99.5% by LCMS).
  • Molecular weight 402.40, chemical formula C23H18N2O5.
  • Available in small-mg packages for research use.
  • Store as powder at -20°C; follow SDS for solution storage and handling.