Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBCO-NHS ester | 1353016-71-3 | 99.5% | 402.40 g/mol | C23H18N2O5 | 100 MG
SDP

Catalog No. 5000204213
Encompass_Preferred

DBCO-NHS ester is an N-hydroxysuccinimide (NHS) activated ester bearing a dibenzocyclooctyne (DBCO) moiety, used as a non-cleavable linker and click-chemistry reagent for bioconjugation and antibody-drug conjugate construction.

  • Non-cleavable ADC linker suitable for bioconjugation.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners.
  • High purity (reported ~99.5%).
  • Physical form: solid; color white to yellow.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Molecular formula C23H18N2O5; molecular weight 402.40 g/mol.

Catalog No. 50-002-04213 Supplier Medchemexpress LLC Supplier No. HY42973100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBCO-NHS ester is an N-hydroxysuccinimide (NHS) activated ester bearing a dibenzocyclooctyne (DBCO) moiety, used as a non-cleavable linker and click-chemistry reagent for bioconjugation and antibody-drug conjugate construction.

  • Non-cleavable ADC linker suitable for bioconjugation.
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners.
  • High purity (reported ~99.5%).
  • Physical form: solid; color white to yellow.
  • Storage: powder -20°C (3 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Molecular formula C23H18N2O5; molecular weight 402.40 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.