Learn More
Medchemexpress LLC DBCO-NHS ester 2 100mg | 1384870-47-6 | 100 MG
SDP
Catalog No. 5000228549
Encompass_Preferred
DBCO-C6-NHS ester is a DBCO-derived N-hydroxysuccinimide (NHS) ester used to introduce a DBCO handle onto amine-containing molecules for copper-free, strain-promoted alkyne-azide cycloaddition (SPAAC) bioconjugation and antibody-drug conjugate (ADC) linker synthesis.
  • Amine-reactive NHS ester for covalent coupling to primary and secondary amines.
  • Facilitates copper-free SPAAC click chemistry with azide-functionalized partners.
  • Suitable for bioconjugation, ADC synthesis, and labeling of proteins or small molecules.
  • Available in multiple sizes; typical laboratory scale is 100 mg.
  • Soluble in DMSO (~14.29 mg/mL); store powder at -20°C for long-term stability.
Catalog No. 50-002-28549 Supplier Medchemexpress LLC Supplier No. HY115524100MG
DBCO-C6-NHS ester is a DBCO-derived N-hydroxysuccinimide (NHS) ester used to introduce a DBCO handle onto amine-containing molecules for copper-free, strain-promoted alkyne-azide cycloaddition (SPAAC) bioconjugation and antibody-drug conjugate (ADC) linker synthesis.
  • Amine-reactive NHS ester for covalent coupling to primary and secondary amines.
  • Facilitates copper-free SPAAC click chemistry with azide-functionalized partners.
  • Suitable for bioconjugation, ADC synthesis, and labeling of proteins or small molecules.
  • Available in multiple sizes; typical laboratory scale is 100 mg.
  • Soluble in DMSO (~14.29 mg/mL); store powder at -20°C for long-term stability.