Learn More
Medchemexpress LLC DBCO-NHS ester 2 1g | 1384870-47-6 | 1 G
SDP
Catalog No. 5000229818
Encompass_Preferred
DBCO-NHS ester 2 is a dibenzoazacyclooctyne (DBCO)-containing N-hydroxysuccinimide (NHS) ester reagent for strain-promoted alkyne-azide cycloaddition (SPAAC). It is used to install a DBCO handle on amine-containing molecules for copper-free click conjugation to azide-functionalized partners and as a cleavable linker in bioconjugation workflows.
  • Amine-reactive NHS ester for efficient coupling to primary amines.
  • DBCO group enables strain-promoted copper-free click chemistry with azides.
  • Suitable for bioconjugation and linker formation in antibody-drug conjugates.
  • High reported purity for reproducible conjugation performance.
  • Supplied as a 1 g quantity for small-scale synthesis and labeling.
Catalog No. 50-002-29818 Supplier Medchemexpress LLC Supplier No. HY1155241G
DBCO-NHS ester 2 is a dibenzoazacyclooctyne (DBCO)-containing N-hydroxysuccinimide (NHS) ester reagent for strain-promoted alkyne-azide cycloaddition (SPAAC). It is used to install a DBCO handle on amine-containing molecules for copper-free click conjugation to azide-functionalized partners and as a cleavable linker in bioconjugation workflows.
  • Amine-reactive NHS ester for efficient coupling to primary amines.
  • DBCO group enables strain-promoted copper-free click chemistry with azides.
  • Suitable for bioconjugation and linker formation in antibody-drug conjugates.
  • High reported purity for reproducible conjugation performance.
  • Supplied as a 1 g quantity for small-scale synthesis and labeling.