Learn More
Medchemexpress LLC DBCO-NHS ester 2 5mg | 1384870-47-6 | 430.45 g/mol | C25H22N2O5 | 5 MG
SDP
Catalog No. 5000227927
Encompass_Preferred
DBCO-NHS ester 2 is an amine-reactive DBCO-derived N-hydroxysuccinimide (NHS) ester used as a click-chemistry linker for strain-promoted alkyne-azide cycloaddition (SPAAC). It enables copper-free conjugation to azide-containing partners and is commonly used in bioconjugation and linker synthesis workflows.
  • Enables copper-free SPAAC with azide-containing partners.
  • Reacts with primary amines via NHS ester chemistry to form stable amide bonds.
  • Useful for linker and probe synthesis in antibody-drug conjugate and labeling applications.
  • Provided in small research-scale masses for convenient handling and formulation.
Catalog No. 50-002-27927 Supplier Medchemexpress LLC Supplier No. HY1155245MG
DBCO-NHS ester 2 is an amine-reactive DBCO-derived N-hydroxysuccinimide (NHS) ester used as a click-chemistry linker for strain-promoted alkyne-azide cycloaddition (SPAAC). It enables copper-free conjugation to azide-containing partners and is commonly used in bioconjugation and linker synthesis workflows.
  • Enables copper-free SPAAC with azide-containing partners.
  • Reacts with primary amines via NHS ester chemistry to form stable amide bonds.
  • Useful for linker and probe synthesis in antibody-drug conjugate and labeling applications.
  • Provided in small research-scale masses for convenient handling and formulation.