Learn More
Cayman Chemical DBCO-NHS ester 250mg
SDP
Catalog No. 502943623
Encompass_Preferred
An azido-reactive click-chemistry reagent; has been conjugated to various anticancer agents for targeting azido-labeled cancer cells in mice
Catalog No. 50-294-3623 Supplier Cayman Chemical Supplier No. 40461250 MG
An azido-reactive click-chemistry reagent; has been conjugated to various anticancer agents for targeting azido-labeled cancer cells in mice