Learn More
Medchemexpress LLC DBCO-NHS ester 3 100mg | 1393350-27-0 | 416.43 | C24H20N2O5 | 100 MG
SDP
Catalog No. 5000228635
Encompass_Preferred
DBCO-NHS ester 3 is an amine-reactive dibenzocyclooctyne (DBCO)-based N-hydroxysuccinimide (NHS) ester used as a clickable linker for constructing antibody-drug conjugates and for labeling amine-containing biomolecules via strain-promoted alkyne-azide cycloaddition. Supplied as a 100 MG solid for research use.
  • Amine-reactive NHS ester enables covalent coupling to primary amines.
  • DBCO moiety supports copper-free click chemistry (SPAAC) with azides.
  • Suitable for antibody-drug conjugate linker synthesis and protein labeling.
  • Provided in small research-scale solid quantities for laboratory workflows.
  • Molecular formula C24H20N2O5; molecular weight 416.43 g/mol.
Catalog No. 50-002-28635 Supplier Medchemexpress LLC Supplier No. HY115545100MG
DBCO-NHS ester 3 is an amine-reactive dibenzocyclooctyne (DBCO)-based N-hydroxysuccinimide (NHS) ester used as a clickable linker for constructing antibody-drug conjugates and for labeling amine-containing biomolecules via strain-promoted alkyne-azide cycloaddition. Supplied as a 100 MG solid for research use.
  • Amine-reactive NHS ester enables covalent coupling to primary amines.
  • DBCO moiety supports copper-free click chemistry (SPAAC) with azides.
  • Suitable for antibody-drug conjugate linker synthesis and protein labeling.
  • Provided in small research-scale solid quantities for laboratory workflows.
  • Molecular formula C24H20N2O5; molecular weight 416.43 g/mol.