Learn More
Medchemexpress LLC DBCO-NHS ester 3 250mg | 1393350-27-0 | 416.43 g·mol⁻1 | C24H20N2O5 | 250 MG
SDP
Catalog No. 5000228636
Encompass_Preferred
DBCO-NHS ester 3 is a dibenzocyclooctyne (DBCO)-N-hydroxysuccinimide (NHS) ester reagent used to functionalize primary amines for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) bioconjugation. It enables installation of a DBCO handle for subsequent conjugation to azide-labeled partners.
  • Installs DBCO onto primary amines for SPAAC (copper-free) click chemistry.
  • Suitable for antibody-drug conjugate linker installation, probe labeling, and general bioconjugation.
  • High purity, typically 95.6%, provided as a solid white to off-white powder.
  • Molecular weight 416.43 g·mol⁻1; formula C24H20N2O5.
  • Available in laboratory-scale 250 MG pack size.
Catalog No. 50-002-28636 Supplier Medchemexpress LLC Supplier No. HY115545250MG
DBCO-NHS ester 3 is a dibenzocyclooctyne (DBCO)-N-hydroxysuccinimide (NHS) ester reagent used to functionalize primary amines for copper-free strain-promoted alkyne-azide cycloaddition (SPAAC) bioconjugation. It enables installation of a DBCO handle for subsequent conjugation to azide-labeled partners.
  • Installs DBCO onto primary amines for SPAAC (copper-free) click chemistry.
  • Suitable for antibody-drug conjugate linker installation, probe labeling, and general bioconjugation.
  • High purity, typically 95.6%, provided as a solid white to off-white powder.
  • Molecular weight 416.43 g·mol⁻1; formula C24H20N2O5.
  • Available in laboratory-scale 250 MG pack size.