Learn More
Medchemexpress LLC DBCO-NHS ester 3 50mg | 1393350-27-0 | 416.43 g/mol | C24H20N2O5 | 50 MG
SDP
Catalog No. 5000228012
Encompass_Preferred
DBCO-NHS ester 3 is an amine-reactive dibenzylcyclooctyne (DBCO) N-hydroxysuccinimide (NHS) ester used for copper-free click chemistry and as a linker in antibody-drug conjugate and bioconjugation workflows. It reacts with primary amines to install a DBCO handle for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners.
  • Amine-reactive NHS ester for efficient conjugation to primary amines.
  • Facilitates copper-free strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Suitable for use as a linker in antibody-drug conjugate and bioconjugation workflows.
  • Supplied as a white to off-white solid, compatible with low-temperature storage.
  • Typical purity around 95.6% and molecular weight 416.43 g/mol support reliable reaction performance.
Catalog No. 50-002-28012 Supplier Medchemexpress LLC Supplier No. HY11554550MG
DBCO-NHS ester 3 is an amine-reactive dibenzylcyclooctyne (DBCO) N-hydroxysuccinimide (NHS) ester used for copper-free click chemistry and as a linker in antibody-drug conjugate and bioconjugation workflows. It reacts with primary amines to install a DBCO handle for strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-functionalized partners.
  • Amine-reactive NHS ester for efficient conjugation to primary amines.
  • Facilitates copper-free strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Suitable for use as a linker in antibody-drug conjugate and bioconjugation workflows.
  • Supplied as a white to off-white solid, compatible with low-temperature storage.
  • Typical purity around 95.6% and molecular weight 416.43 g/mol support reliable reaction performance.