Learn More
Cayman Chemical DBCO-NHS ester, Purity: ≥95%, CAS: 1353016-71-3, 50 mg
SDP
Catalog No. 502942809
Encompass_Preferred
An azido-reactive click-chemistry reagent; has been conjugated to various anticancer agents for targeting azido-labeled cancer cells in mice
Catalog No. 50-294-2809 Supplier Cayman Chemical Supplier No. 4046150 MG
An azido-reactive click-chemistry reagent; has been conjugated to various anticancer agents for targeting azido-labeled cancer cells in mice