Learn More
Medchemexpress LLC DBCO-(PEG)3-VC-PAB-MMAE | 2754384-60-4 | 99.60% | 1613.97 | 100 UG
SDP
Catalog No. 5000343278
Encompass_Preferred
DBCO-(PEG)3-VC-PAB-MMAE is an agent-linker conjugate for ADC, formed by conjugating Monomethyl auristatin E (HY-15162) to a DBCO-(PEG)3-vc-PAB linker. It can be used for cancer research and functions as a click chemistry reagent, containing a DBCO group that undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • Agent-linker conjugate for ADC
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC with azide groups
  • For research use only
Catalog No. 50-003-43278 Supplier Medchemexpress LLC Supplier No. HY1110121MG 10
DBCO-(PEG)3-VC-PAB-MMAE is an agent-linker conjugate for ADC, formed by conjugating Monomethyl auristatin E (HY-15162) to a DBCO-(PEG)3-vc-PAB linker. It can be used for cancer research and functions as a click chemistry reagent, containing a DBCO group that undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups.
  • Agent-linker conjugate for ADC
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC with azide groups
  • For research use only