Learn More
Medchemexpress LLC DBCO-PEG1-NH-Boc | 2364591-77-3 | C28H33N3O5 | 50 MG
SDP
Catalog No. 5000353037
Encompass_Preferred
DBCO-PEG1-NH-Boc is a PEG linker featuring a DBCO group and a Boc-protected amine. The DBCO component enables copper-free Click Chemistry reactions with azides, while the Boc protecting group can be removed under acidic conditions for further reactions. This compound can be used in the synthesis of PROTACs and as a click chemistry reagent.
  • Features a DBCO group and a Boc-protected amine
  • Facilitates copper-free Click Chemistry reactions with azides
  • Boc protecting group can be removed under acidic conditions
  • Categorized as a PEG-based PROTAC linker
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups
Catalog No. 50-003-53037 Supplier Medchemexpress LLC Supplier No. HY14028950MG
DBCO-PEG1-NH-Boc is a PEG linker featuring a DBCO group and a Boc-protected amine. The DBCO component enables copper-free Click Chemistry reactions with azides, while the Boc protecting group can be removed under acidic conditions for further reactions. This compound can be used in the synthesis of PROTACs and as a click chemistry reagent.
  • Features a DBCO group and a Boc-protected amine
  • Facilitates copper-free Click Chemistry reactions with azides
  • Boc protecting group can be removed under acidic conditions
  • Categorized as a PEG-based PROTAC linker
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups