Learn More
eMolecules​ 2228857-34-7 | DBCO-PEG1-NHS ester | Broadpharm517.538 | C28H27N3O7 | 98.000 | O=C(CCC(=O)N1Cc2ccccc2C#Cc2ccccc12)NCCOCCC(=O)ON1C(=O)CCC1=O | 100mg | 393649427
Catalog No. NC1698324 Shop All eMolecules Inc. Products
Encompass_Preferred
DBCO-PEG1-NHS ester | Broadpharm | 2228857-34-7517.538 | C28H27N3O7 | 98.000 | O=C(CCC(=O)N1Cc2ccccc2C#Cc2ccccc12)NCCOCCC(=O)ON1C(=O)CCC1=O | 100mg | 393649427
Catalog No. NC1698324 Supplier eMolecules​ Supplier No. 393649427
DBCO-PEG1-NHS ester | Broadpharm | 2228857-34-7517.538 | C28H27N3O7 | 98.000 | O=C(CCC(=O)N1Cc2ccccc2C#Cc2ccccc12)NCCOCCC(=O)ON1C(=O)CCC1=O | 100mg | 393649427