Learn More
Medchemexpress LLC DBCO-PEG1-NHS ester | 2228857-34-7 | 95.0% | C28H27N3O7 | 50 MG
SDP
Catalog No. 5000357247
Encompass_Preferred
DBCO-PEG1-NHS ester is a PEG-based PROTAC linker. It is also a click chemistry reagent that contains a DBCO group, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC
  • Applicable in cancer research and targeted therapy
Catalog No. 50-003-57247 Supplier Medchemexpress LLC Supplier No. HY14027150MG
DBCO-PEG1-NHS ester is a PEG-based PROTAC linker. It is also a click chemistry reagent that contains a DBCO group, enabling strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC
  • Applicable in cancer research and targeted therapy