Learn More
Medchemexpress LLC DBCO-PEG12-maleimide | 00-00-0 | 98.0% | C52H74N4O17 | 10MG
SDP
Catalog No. 5000259481
Encompass_Preferred
DBCO-PEG12-maleimide is a PEG12-based bifunctional linker for bioconjugation and PROTAC synthesis. It combines a dibenzocyclooctyne (DBCO) moiety for strain-promoted alkyne-azide cycloaddition and a maleimide group for selective thiol conjugation, providing a flexible, hydrophilic spacer for assembling complex conjugates. Supplied as a high-purity reagent for research use.
  • Enables copper-free click chemistry via strain-promoted alkyne-azide cycloaddition.
  • Provides thiol-selective conjugation through a maleimide reactive handle.
  • Offers a PEG12 spacer to increase solubility and reduce steric hindrance.
  • High purity suitable for research applications.
  • Stable when stored under recommended conditions.
Catalog No. 50-002-59481 Supplier Medchemexpress LLC Supplier No. HY13337410MG
DBCO-PEG12-maleimide is a PEG12-based bifunctional linker for bioconjugation and PROTAC synthesis. It combines a dibenzocyclooctyne (DBCO) moiety for strain-promoted alkyne-azide cycloaddition and a maleimide group for selective thiol conjugation, providing a flexible, hydrophilic spacer for assembling complex conjugates. Supplied as a high-purity reagent for research use.
  • Enables copper-free click chemistry via strain-promoted alkyne-azide cycloaddition.
  • Provides thiol-selective conjugation through a maleimide reactive handle.
  • Offers a PEG12 spacer to increase solubility and reduce steric hindrance.
  • High purity suitable for research applications.
  • Stable when stored under recommended conditions.