Learn More
Medchemexpress LLC DBCO-PEG12-maleimide | 98.0% | 1027.16 g/mol | C52H74N4O17 | 5 MG
SDP
Catalog No. 5000262406
Encompass_Preferred
DBCO-PEG12-maleimide is a bifunctional polyethylene glycol (PEG12) linker bearing a dibenzocyclooctyne (DBCO) group for strain-promoted alkyne-azide cycloaddition and a terminal maleimide for thiol-specific conjugation. It is designed for catalyst-free click chemistry and selective cysteine conjugation in bioconjugation workflows, linker synthesis, and chemical biology applications.
  • Enables copper-free click (SPAAC) reactions with azide-functionalized partners.
  • Reacts selectively with thiols via the maleimide to form stable thioether bonds.
  • PEG12 spacer improves solubility and provides flexible linker length.
  • High purity suitable for sensitive bioconjugation applications.
  • Stable when stored dry at recommended low temperatures.
Catalog No. 50-002-62406 Supplier Medchemexpress LLC Supplier No. HY1333745MG
DBCO-PEG12-maleimide is a bifunctional polyethylene glycol (PEG12) linker bearing a dibenzocyclooctyne (DBCO) group for strain-promoted alkyne-azide cycloaddition and a terminal maleimide for thiol-specific conjugation. It is designed for catalyst-free click chemistry and selective cysteine conjugation in bioconjugation workflows, linker synthesis, and chemical biology applications.
  • Enables copper-free click (SPAAC) reactions with azide-functionalized partners.
  • Reacts selectively with thiols via the maleimide to form stable thioether bonds.
  • PEG12 spacer improves solubility and provides flexible linker length.
  • High purity suitable for sensitive bioconjugation applications.
  • Stable when stored dry at recommended low temperatures.