Learn More
Medchemexpress LLC DBCO-PEG12-NHS ester | 2093934-94-0 | 96.1% | C50H71N3O18 | 25 MG
SDP
Catalog No. 5000357352
Encompass_Preferred
DBCO-PEG12-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups. PROTACs utilize the ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Features a DBCO group for SPAAC
  • Facilitates strain-promoted alkyne-azide cycloaddition with azide groups
  • PROTACs utilize the ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-57352 Supplier Medchemexpress LLC Supplier No. 25MGHY140274
DBCO-PEG12-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide groups. PROTACs utilize the ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Features a DBCO group for SPAAC
  • Facilitates strain-promoted alkyne-azide cycloaddition with azide groups
  • PROTACs utilize the ubiquitin-proteasome system
  • Selectively degrades target proteins