Learn More
Medchemexpress LLC DBCO-PEG12-NHS ester | 2093934-94-0 | 96.12% | C50H71N3O18 | 100 MG
SDP
Catalog No. 5000356829
Encompass_Preferred
DBCO-PEG12-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, utilizing a DBCO group to facilitate strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Enables synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Utilizes DBCO group for SPAAC with azide-containing molecules
  • Supports selective degradation of target proteins
Catalog No. 50-003-56829 Supplier Medchemexpress LLC Supplier No. HY140274100MG
DBCO-PEG12-NHS ester is a PEG-based PROTAC linker designed for the synthesis of PROTACs. It functions as a click chemistry reagent, utilizing a DBCO group to facilitate strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs leverage the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Enables synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Utilizes DBCO group for SPAAC with azide-containing molecules
  • Supports selective degradation of target proteins