Learn More
Medchemexpress LLC DBCO-PEG12-NHS ester | 2093934-94-0 | 98.7% | C50H71N3O18 | 50 MG
SDP
Catalog No. 5000353098
Encompass_Preferred
DBCO-PEG12-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC with Azide groups
  • Oil appearance
  • Colorless to light yellow color
Catalog No. 50-003-53098 Supplier Medchemexpress LLC Supplier No. HY14027450MG
DBCO-PEG12-NHS ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent, containing a DBCO group that can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing Azide groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains a DBCO group for SPAAC with Azide groups
  • Oil appearance
  • Colorless to light yellow color