Learn More
Broadpharm DBCO-PEG12-NHS ester| 98% 50mg CAS#2093934-94-0 C50H71N3O18 MW:1002.1
Catalog No. NC2208306
Encompass
DBCO-PEG12-NHS ester| 98% 50mg CAS#2093934-94-0 C50H71N3O18 MW:1002.1
This item is not returnable. View return policy
Catalog No. NC2208306 Supplier Broadpharm Supplier No. BP2414950MG
DBCO-PEG12-NHS ester| 98% 50mg CAS#2093934-94-0 C50H71N3O18 MW:1002.1