Learn More
Medchemexpress LLC DBCO-PEG2-acid | 2304558-25-4 | ≥95.0% | C26H28N2O6 | 250 MG
SDP
Catalog No. 5000352863
Encompass_Preferred
DBCO-PEG2-acid is a PEG-based PROTAC linker designed for synthesizing PROTACs. It acts as a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs typically comprise two distinct ligands connected by a linker, where one targets an E3 ubiquitin ligase and the other targets the protein of interest, enabling selective degradation through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Incorporates a DBCO group
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Suitable for laboratory chemical applications
Catalog No. 50-003-52863 Supplier Medchemexpress LLC Supplier No. 250MGHY140266
DBCO-PEG2-acid is a PEG-based PROTAC linker designed for synthesizing PROTACs. It acts as a click chemistry reagent, featuring a DBCO group that facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) with azide-containing molecules. PROTACs typically comprise two distinct ligands connected by a linker, where one targets an E3 ubiquitin ligase and the other targets the protein of interest, enabling selective degradation through the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Incorporates a DBCO group
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Used in the synthesis of PROTACs
  • Leverages the intracellular ubiquitin-proteasome system
  • Suitable for laboratory chemical applications