Learn More
Medchemexpress LLC DBCO-PEG2-DBCO | 2639395-48-3 | C44H42N4O6 | 25 MG
SDP
Catalog No. 5000375979
Encompass_Preferred
DBCO-PEG2-DBCO is a click chemistry reagent that contains a DBCO group. It functions as a PEG linker with two terminal DBCO groups. These DBCO groups are frequently used for copper-free Click Chemistry reactions due to their strain-promoted high energy. The hydrophilic PEG chain in DBCO-PEG2-DBCO enhances its water solubility. This product is reagent grade and intended for research use only.
  • Contains a DBCO group
  • Functions as a PEG linker with two terminal DBCO groups
  • Commonly utilized in copper-free click chemistry reactions
  • Exhibits strain-promoted high energy
  • Hydrophilic PEG chain enhances water solubility
  • Reagent grade
  • Intended for research use only
Catalog No. 50-003-75979 Supplier Medchemexpress LLC Supplier No. 25MGHY151834
DBCO-PEG2-DBCO is a click chemistry reagent that contains a DBCO group. It functions as a PEG linker with two terminal DBCO groups. These DBCO groups are frequently used for copper-free Click Chemistry reactions due to their strain-promoted high energy. The hydrophilic PEG chain in DBCO-PEG2-DBCO enhances its water solubility. This product is reagent grade and intended for research use only.
  • Contains a DBCO group
  • Functions as a PEG linker with two terminal DBCO groups
  • Commonly utilized in copper-free click chemistry reactions
  • Exhibits strain-promoted high energy
  • Hydrophilic PEG chain enhances water solubility
  • Reagent grade
  • Intended for research use only