Learn More
Medchemexpress LLC Dbco-peg2-dbco | 2639395-48-3 | C44H42N4O6 | 5 MG
SDP
Catalog No. 5000374015
Encompass_Preferred
DBCO-PEG2-DBCO is a click chemistry reagent containing a DBCO group. It is a PEG linker with two terminal DBCO groups, which are commonly used for copper-free Click Chemistry reactions due to their strain-promoted high energy. The hydrophilic PEG chain increases water solubility. This product is reagent grade and for research use only.
  • Storage conditions:
  • Powder: -20°C for 3 years
  • In solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-74015 Supplier Medchemexpress LLC Supplier No. HY1518345MG
DBCO-PEG2-DBCO is a click chemistry reagent containing a DBCO group. It is a PEG linker with two terminal DBCO groups, which are commonly used for copper-free Click Chemistry reactions due to their strain-promoted high energy. The hydrophilic PEG chain increases water solubility. This product is reagent grade and for research use only.
  • Storage conditions:
  • Powder: -20°C for 3 years
  • In solvent: -80°C for 6 months, -20°C for 1 month